siRNA Details
Gene Symbol: SNAI3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024399.1 | 4 | 893 | |||
GenBank mRNA | AY203928.1 | - | 1 | 2026 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SNAI3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024399.1 | 4 | 893 | |||
GenBank mRNA | AY203928.1 | - | 1 | 2026 |