siRNA Details
Gene Symbol: SNAI3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024402.2 | 4 | 549 | |||
GenBank mRNA | AK094837.1 | - | 1 | 230 | ||
BX640980.1 | - | 4 | 569 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SNAI3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024402.2 | 4 | 549 | |||
GenBank mRNA | AK094837.1 | - | 1 | 230 | ||
BX640980.1 | - | 4 | 569 |