siRNA Details
Gene Symbol: SNAI3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024402.2 | 4 | 882 | |||
GenBank mRNA | AK094837.1 | - | 1 | 563 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA254069.1 | - | 1 | 563 | Chr. 16 - 88673396 - 88673986 [+] on Build GRCh38 |
