siRNA Details
Gene Symbol: LOC100505915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125434.1 | 4 | 1458 | |||
GenBank mRNA | AK125313.1 | - | 4 | 1458 | ||
BC017937.1 | - | 1 | 186 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100505915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125434.1 | 4 | 1458 | |||
GenBank mRNA | AK125313.1 | - | 4 | 1458 | ||
BC017937.1 | - | 1 | 186 |