siRNA Details
Gene Symbol: LOC101929634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135293.1 | 2 | 3053 | |||
GenBank mRNA | AK128797.1 | - | 2 | 3050 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929634 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135293.1 | 2 | 3053 | |||
GenBank mRNA | AK128797.1 | - | 2 | 3050 |