siRNA Details
Gene Symbol: ADPGK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040107.1 | 1 | 1191 | |||
GenBank mRNA | AK127877.1 | - | 1 | 1193 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ADPGK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040107.1 | 1 | 1191 | |||
GenBank mRNA | AK127877.1 | - | 1 | 1193 |