siRNA Details
Gene Symbol: ADPGK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040107.1 | 2 | 2454 | |||
GenBank mRNA | AK127877.1 | - | 2 | 2456 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK127230.1 | - | 379 | Chr. - - [-] on Build |
