siRNA Details
Gene Symbol: LOC101927450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109771.1 | 4 | 576 | |||
GenBank mRNA | BC042427.1 | - | 4 | 576 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG719985.1 | - | 4 | 581 | Chr. 9 - 79137037 - 79145666 [-] on Build GRCh38 |
