siRNA Details
Gene Symbol: WFDC21P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KJ020271.1 | - | 2, 3 | 211 | ||
KJ020272.1 | - | 1, 2 | 191 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WFDC21P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KJ020271.1 | - | 2, 3 | 211 | ||
KJ020272.1 | - | 1, 2 | 191 |