siRNA Details
Gene Symbol: RBFADN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103445.2 | 4 | 643 | |||
GenBank mRNA | CN288269.1 | - | 1 | 331 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RBFADN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103445.2 | 4 | 643 | |||
GenBank mRNA | CN288269.1 | - | 1 | 331 |