siRNA Details
Gene Symbol: C16orf97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242473.1 | NP_001229402.1 | 6 | 419 | ||
XM_011523081.1 | XP_011521383.1 | 8 | 681 | |||
GenBank mRNA | BX096212.1 | - | 6 | 419 |
