siRNA Details
Gene Symbol: LOC100507534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110649.1 | 2 | 720 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG708079.1 | - | 2 | 725 | Chr. 16 - 47849903 - 47887131 [-] on Build GRCh38 |
