siRNA Details
Gene Symbol: VPS9D1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036480.1 | 4 | 1174 | |||
GenBank mRNA | AK096286.1 | - | 4 | 1175 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VPS9D1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036480.1 | 4 | 1174 | |||
GenBank mRNA | AK096286.1 | - | 4 | 1175 |