siRNA Details
Gene Symbol: VPS9D1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036480.1 | 3 | 905 | |||
GenBank mRNA | AK096286.1 | - | 3 | 906 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM909691.1 | - | 679 | Chr. - - [-] on Build |
