siRNA Details
Gene Symbol: LOC101928035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104657.1 | 1, 2 | 171 | |||
GenBank mRNA | BC043527.1 | - | 1, 2 | 171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI826257.1 | - | 1, 2 | 171 | Chr. 16 - 74192525 - 74215521 [-] on Build GRCh38 |
