siRNA Details
Gene Symbol: LINC00558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047488.1 | 4, 5 | 441 | |||
GenBank mRNA | BC041877.1 | - | 4, 5 | 441 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI465050.1 | - | 4, 5 | 445 | Chr. 13 - 53815417 - 53873533 [+] on Build GRCh38 |
