siRNA Details
Gene Symbol: DLGAP1-AS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036489.1 | 1 | 107 | |||
GenBank mRNA | AK098180.1 | - | 1 | 76 | ||
DA301744.1 | - | 1 | 109 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB205872.1 | - | 1 | 76 | Chr. 18 - 4264633 - 4293412 [+] on Build GRCh38 |
