siRNA Details
Gene Symbol: LOC101929144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110745.1 | 5 | 745 | |||
GenBank mRNA | BC037873.1 | - | 5 | 750 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110745.1 | 5 | 745 | |||
GenBank mRNA | BC037873.1 | - | 5 | 750 |