siRNA Details
Gene Symbol: LOC101929144 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110745.1 | 2, 3 | 455 | |||
GenBank mRNA | BC037873.1 | - | 2, 3 | 460 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI465107.1 | - | 2, 3 | 457 | Chr. 19 - 23058464 - 23061265 [-] on Build GRCh38 |
