siRNA Details
Gene Symbol: LINC00838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038932.1 | 3 | 2354 | |||
GenBank mRNA | BC042457.1 | - | 3 | 2332 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038932.1 | 3 | 2354 | |||
GenBank mRNA | BC042457.1 | - | 3 | 2332 |