siRNA Details
Gene Symbol: LINC00838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038932.1 | 1 | 481 | |||
GenBank mRNA | BC042457.1 | - | 1 | 459 | ||
DB078335.1 | - | 1 | 481 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038932.1 | 1 | 481 | |||
GenBank mRNA | BC042457.1 | - | 1 | 459 | ||
DB078335.1 | - | 1 | 481 |