siRNA Details
Gene Symbol: LOC101928682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057367.1 | - | 1 | 432 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB509679.1 | - | 1 | 32 | Chr. 16 - 87290726 - 87292038 [-] on Build GRCh38 |
