siRNA Details
Gene Symbol: LOXL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040066.1 | 4 | 901 | |||
NR_040067.1 | 4 | 779 | ||||
NR_040068.1 | 3 | 716 | ||||
NR_040069.1 | 3 | 594 | ||||
NR_040070.1 | 3 | 594 | ||||
GenBank mRNA | AK056376.1 | - | 3 | 716 | ||
AK056885.1 | - | 4 | 901 | |||
BC017577.1 | - | 1 | 330 | |||
BX366015.2 | - | 3 | 595 | |||
DA021396.1 | - | 3 | 549 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX383692.2 | - | 3 | 594 | Chr. 15 - 73918829 - 73927942 [-] on Build GRCh38 |
