siRNA Details
Gene Symbol: LOXL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040066.1 | 4 | 709 | |||
NR_040067.1 | 4 | 587 | ||||
NR_040068.1 | 3 | 524 | ||||
NR_040069.1 | 3 | 402 | ||||
NR_040070.1 | 3 | 402 | ||||
GenBank mRNA | AK056376.1 | - | 3 | 524 | ||
AK056885.1 | - | 4 | 709 | |||
BC017577.1 | - | 1 | 138 | |||
BX366015.2 | - | 3 | 403 | |||
DA021396.1 | - | 3 | 357 | |||
DA038997.1 | - | 4 | 496 |
