siRNA Details
Gene Symbol: LOXL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040066.1 | 3 | 430 | |||
NR_040067.1 | 3 | 430 | ||||
NR_040068.1 | 2 | 245 | ||||
NR_040069.1 | 2 | 245 | ||||
NR_040070.1 | 2 | 245 | ||||
GenBank mRNA | AK056376.1 | - | 2 | 245 | ||
AK056885.1 | - | 3 | 430 | |||
BX355383.2 | - | 2 | 244 | |||
BX366015.2 | - | 2 | 246 | |||
DA021396.1 | - | 2 | 200 | |||
DA038997.1 | - | 3 | 339 |
