siRNA Details
Gene Symbol: LOC102724927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120311.1 | 2 | 663 | |||
GenBank mRNA | BC012429.1 | - | 2 | 663 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF037667.1 | - | 2 | 635 | Chr. 16 - 3948189 - 3950422 [-] on Build GRCh38 |
