siRNA Details
Gene Symbol: LOC102724927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120311.1 | 2 | 310 | |||
GenBank mRNA | BC012429.1 | - | 2 | 310 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA435098.1 | - | 1 | 470 | Chr. 16 - 3948523 - 3949158 [-] on Build GRCh38 |
