siRNA Details
Gene Symbol: TMEM220-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073455.1 | 3 | 209 | |||
NR_073456.1 | 2 | 113 | ||||
GenBank mRNA | BQ694074.1 | - | 2 | 79 | ||
BQ694934.1 | - | 2 | 67 | |||
DA867863.1 | - | 3 | 190 |
