siRNA Details
Gene Symbol: TMEM220-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073455.1 | 5 | 574 | |||
NR_073456.1 | 4 | 478 | ||||
GenBank mRNA | BQ694074.1 | - | 4 | 444 | ||
BQ694934.1 | - | 4 | 432 | |||
DA747858.1 | - | 3 | 444 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI218490.1 | - | 3 | 422 | Chr. 17 - 10729797 - 10766921 [+] on Build GRCh38 |
