siRNA Details
Gene Symbol: HID1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110878.1 | 2 | 254 | |||
GenBank mRNA | CD515974.1 | - | 2 | 239 | ||
HY081987.1 | - | 2 | 254 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HID1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110878.1 | 2 | 254 | |||
GenBank mRNA | CD515974.1 | - | 2 | 239 | ||
HY081987.1 | - | 2 | 254 |