siRNA Details
Gene Symbol: LOC102724532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135480.1 | 2 | 162 | |||
GenBank mRNA | HY017353.1 | - | 2 | 163 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK127217.1 | - | 1 | 1878 | Chr. 17 - 47891854 - 47895792 [-] on Build GRCh38 |
