siRNA Details
Gene Symbol: TSPOAP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038410.1 | 5 | 1553 | |||
NR_038411.1 | 4 | 1348 | ||||
NR_038412.1 | 4 | 1339 | ||||
NR_038413.1 | 4 | 1271 | ||||
NR_038414.1 | 4 | 1521 | ||||
NR_038415.1 | 3 | 1457 | ||||
NR_038416.1 | 3 | 1448 | ||||
NR_038417.1 | 4 | 1320 | ||||
NR_038418.1 | 4 | 1311 | ||||
GenBank mRNA | AK000271.1 | - | 4 | 1415 | ||
AK090885.1 | - | 5 | 1552 | |||
BC032475.1 | - | 3 | 1301 | |||
BC050403.1 | - | 3 | 1301 |
