siRNA Details
Gene Symbol: TSPOAP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038410.1 | 3 | 558 | |||
NR_038411.1 | 2 | 344 | ||||
NR_038412.1 | 2 | 344 | ||||
NR_038413.1 | 2 | 267 | ||||
NR_038414.1 | 1 | 453 | ||||
NR_038415.1 | 1 | 453 | ||||
NR_038416.1 | 1 | 453 | ||||
NR_038417.1 | 2 | 316 | ||||
NR_038418.1 | 2 | 316 | ||||
GenBank mRNA | AK000271.1 | - | 1 | 347 | ||
AK090885.1 | - | 3 | 558 | |||
AK311208.1 | - | 2 | 277 | |||
AK311311.1 | - | 2 | 344 | |||
BC032475.1 | - | 1 | 297 | |||
BC050403.1 | - | 1 | 306 | |||
BI820069.1 | - | 2 | 267 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI839033.1 | - | 1 | 425 | Chr. 17 - 58337244 - 58415766 [+] on Build GRCh38 |
