siRNA Details
Gene Symbol: TSPOAP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038410.1 | 3 | 422 | |||
NR_038411.1 | 2 | 208 | ||||
NR_038412.1 | 2 | 208 | ||||
NR_038413.1 | 2 | 131 | ||||
NR_038414.1 | 1 | 317 | ||||
NR_038415.1 | 1 | 317 | ||||
NR_038416.1 | 1 | 317 | ||||
NR_038417.1 | 2 | 180 | ||||
NR_038418.1 | 2 | 180 | ||||
GenBank mRNA | AK000271.1 | - | 1 | 211 | ||
AK311208.1 | - | 2 | 141 | |||
AK311306.1 | - | 2 | 179 | |||
AK311311.1 | - | 2 | 208 | |||
BC032475.1 | - | 1 | 161 | |||
BC050403.1 | - | 1 | 170 | |||
BI820069.1 | - | 2 | 131 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI839033.1 | - | 1 | 289 | Chr. 17 - 58337244 - 58415766 [+] on Build GRCh38 |
