siRNA Details
Gene Symbol: LOC101927168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134645.1 | 2 | 210 | |||
GenBank mRNA | BG202214.1 | - | 1 | 71 | ||
BX097886.1 | - | 2 | 210 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134645.1 | 2 | 210 | |||
GenBank mRNA | BG202214.1 | - | 1 | 71 | ||
BX097886.1 | - | 2 | 210 |