siRNA Details
Gene Symbol: LOC101927168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134645.1 | 2 | 344 | |||
GenBank mRNA | BG202214.1 | - | 1 | 205 | ||
BX097886.1 | - | 2 | 344 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB335425.1 | - | 1 | 14 | Chr. 18 - 6728475 - 6728981 [-] on Build GRCh38 |
