siRNA Details
Gene Symbol: LOC105372069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134585.1 | 11 | ||||
GenBank mRNA | BC037214.1 | - | 11 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105372069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134585.1 | 11 | ||||
GenBank mRNA | BC037214.1 | - | 11 |