siRNA Details
Gene Symbol: RNF157-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040017.1 | 8 | 1966 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB508842.1 | - | 1 | 233 | Chr. 17 - 76154204 - 76154647 [+] on Build GRCh38 |
