siRNA Details
Gene Symbol: RNF157-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040017.1 | 6, 7 | 923 | |||
GenBank mRNA | AK093500.1 | - | 6, 7 | 923 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNF157-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040017.1 | 6, 7 | 923 | |||
GenBank mRNA | AK093500.1 | - | 6, 7 | 923 |