siRNA Details
Gene Symbol: LINC01140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026985.1 | 4 | 1020 | |||
NR_026986.1 | 3 | 820 | ||||
NR_026987.1 | 3 | 813 | ||||
NR_026988.1 | 3 | 716 | ||||
GenBank mRNA | AL833352.1 | - | 4 | 1044 | ||
BC030523.1 | - | 2 | 857 | |||
BC041867.1 | - | 4 | 991 |
