siRNA Details
Gene Symbol: LOC105372397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134907.1 | 2 | 205 | |||
GenBank mRNA | BM716908.1 | - | 2 | 206 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105372397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134907.1 | 2 | 205 | |||
GenBank mRNA | BM716908.1 | - | 2 | 206 |