siRNA Details
Gene Symbol: ZNF582-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037159.1 | 1 | 342 | |||
NR_037160.1 | 1 | 342 | ||||
NR_037161.1 | 1 | 342 | ||||
GenBank mRNA | AW160702.1 | - | 1 | 343 | ||
BC024249.1 | - | 1 | 276 |
