siRNA Details
Gene Symbol: ZNF582-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037159.1 | 4 | 1547 | |||
NR_037160.1 | 2 | 1550 | ||||
NR_037161.1 | 4 | 1200 | ||||
GenBank mRNA | BC024249.1 | - | 2 | 1484 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF582-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037159.1 | 4 | 1547 | |||
NR_037160.1 | 2 | 1550 | ||||
NR_037161.1 | 4 | 1200 | ||||
GenBank mRNA | BC024249.1 | - | 2 | 1484 |