siRNA Details
Gene Symbol: THEG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278575.1 | NP_001265504.1 | 3 | 432 | ||
NM_001278576.1 | NP_001265505.1 | 4 | 556 | |||
NM_001278577.1 | NP_001265506.1 | 4 | 567 | |||
NM_001278578.1 | NP_001265507.1 | 3 | 425 | |||
XM_011526302.2 | XP_011524604.1 | 3 | 2211 | |||
XM_011526303.2 | XP_011524605.1 | 3 | 1036 | |||
XM_017026126.1 | XP_016881615.1 | 3 | 2547 | |||
XM_017026127.1 | XP_016881616.1 | 3 | 2006 | |||
XM_017026128.1 | XP_016881617.1 | 3 | 2204 | |||
XM_017026129.1 | XP_016881618.1 | 3 | 1043 | |||
XM_017026130.1 | XP_016881619.1 | 3 | 768 | |||
XM_017026131.1 | XP_016881620.1 | 4 | 374 | |||
XM_017026132.1 | XP_016881621.1 | 3 | 567 | |||
XM_017026133.1 | XP_016881622.1 | 2 | 235 | |||
GenBank mRNA | BC043286.1 | - | 4 | 495 | ||
DB054421.1 | - | 3 | 419 | |||
HY013681.1 | - | 3 | 433 |
