siRNA Details
Gene Symbol: ZNF571-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038247.1 | 5 | 1119 | |||
NR_038248.1 | 6 | 1030 | ||||
NR_038249.1 | 4 | 855 | ||||
NR_038250.1 | 3 | 796 | ||||
GenBank mRNA | BC043284.1 | - | 6 | 1033 | ||
BC044917.1 | - | 4 | 857 | |||
BC110871.1 | - | 4 | 682 | |||
BX647800.1 | - | 3 | 786 |
