siRNA Details
Gene Symbol: ZNF571-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038247.1 | 5 | 789 | |||
NR_038248.1 | 6 | 700 | ||||
NR_038249.1 | 4 | 525 | ||||
NR_038250.1 | 3 | 466 | ||||
GenBank mRNA | BC042488.1 | - | 5 | 791 | ||
BC043284.1 | - | 6 | 703 | |||
BC044917.1 | - | 4 | 527 | |||
BC110871.1 | - | 4 | 352 | |||
BX647800.1 | - | 3 | 456 |
