siRNA Details
Gene Symbol: LOC644554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040013.1 | 5 | 830 | |||
GenBank mRNA | AK092337.1 | - | 4 | 630 | ||
AK309685.1 | - | 5 | 830 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX954907.1 | - | 4 | 551 | Chr. 19 - 37817595 - 37826474 [+] on Build GRCh38 |
