siRNA Details
Gene Symbol: LDLRAD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040031.1 | 4 | 1057 | |||
GenBank mRNA | AK098298.1 | - | 4 | 1003 | ||
BC042085.1 | - | 4 | 1042 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LDLRAD4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040031.1 | 4 | 1057 | |||
GenBank mRNA | AK098298.1 | - | 4 | 1003 | ||
BC042085.1 | - | 4 | 1042 |