siRNA Details
Gene Symbol: CARD8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040599.1 | 2 | 1155 | |||
GenBank mRNA | AK125548.1 | - | 2 | 1155 | ||
BC008448.1 | - | 2 | 1134 | |||
BC060888.1 | - | 2 | 1141 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CARD8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040599.1 | 2 | 1155 | |||
GenBank mRNA | AK125548.1 | - | 2 | 1155 | ||
BC008448.1 | - | 2 | 1134 | |||
BC060888.1 | - | 2 | 1141 |