siRNA Details
Gene Symbol: NAPA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038452.1 | 1, 2 | 244 | |||
GenBank mRNA | AK057858.1 | - | 1, 2 | 244 | ||
BG035979.1 | - | 1, 2 | 218 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NAPA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038452.1 | 1, 2 | 244 | |||
GenBank mRNA | AK057858.1 | - | 1, 2 | 244 | ||
BG035979.1 | - | 1, 2 | 218 |